Gene Transcripts

Gene Symbol
OR4A5
Entrez Gene ID
81318
Gene Name
olfactory receptor family 4 subfamily A member 5
Gene Aliases
OR11-111
Chromosome Location
Chr. 11: 54706832 - 54707902 on Build GRCh38
UniGene ID
Hs.554531
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005272.3 NP_001005272.3 1 916
GenBank mRNA BC140750.1 - 1 916

View Full Product Details