Gene Transcripts

Gene Symbol
UNC93B1
Entrez Gene ID
81622
Gene Name
unc-93 homolog B1 (C. elegans)
Gene Aliases
IIAE1, UNC93, UNC93B, Unc-93B1
Chromosome Location
Chr. 11: 67991100 - 68004125 on Build GRCh38
UniGene ID
Hs.502989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030930.3 NP_112192.2 3 427
XM_011545290.1 XP_011543592.1 1 36
XM_011545291.2 XP_011543593.1 2 249
GenBank mRNA AJ271326.1 - 3 386
AJ422142.1 - 3 395
AY007125.1 - 2 214
BC025669.1 - 3 373
BC033623.1 - 3 425
BC092472.1 - 3 395 30348121
BC101568.1 - 3 371 8069074
BC105104.1 - 3 371 8144107
HY081711.1 - 3 427

View Full Product Details