Gene Transcripts
- Gene Symbol
- HORMAD1
- Entrez Gene ID
- 84072
- Gene Name
- HORMA domain containing 1
- Gene Aliases
- CT46, NOHMA
- Chromosome Location
- Chr. 1: 150698059 - 150720910 on Build GRCh38
- UniGene ID
- Hs.298312
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199829.1 |
NP_001186758.1
|
9, 10 |
890 |
|
NM_032132.4 |
NP_115508.2
|
10, 11 |
911 |
|
XM_006711570.2 |
XP_006711633.1
|
9, 10 |
805 |
|
XM_006711571.2 |
XP_006711634.1
|
6, 7 |
667 |
|
XM_011510053.2 |
XP_011508355.1
|
10, 11 |
979 |
|
XM_011510054.2 |
XP_011508356.1
|
10, 11 |
933 |
|
XM_011510055.2 |
XP_011508357.1
|
9, 10 |
729 |
|
XM_011510056.2 |
XP_011508358.1
|
5, 6 |
511 |
|
GenBank mRNA |
AK097390.1 |
- |
10, 11 |
899 |
|
AL136755.2 |
- |
9, 10 |
887 |
|
AY626344.1 |
- |
9, 10 |
887 |
|
BC047406.1 |
- |
10, 11 |
899 |
5298160 |
View Full Product Details