Gene Transcripts

Gene Symbol
HVCN1
Entrez Gene ID
84329
Gene Name
hydrogen voltage gated channel 1
Gene Aliases
HV1, VSOP
Chromosome Location
Chr. 12: 110648686 - 110704945 on Build GRCh38
UniGene ID
Hs.211511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040107.1 NP_001035196.1 8 995
NM_001256413.1 NP_001243342.1 7 955
NM_032369.3 NP_115745.2 8 986
XM_005253948.2 XP_005254005.1 8 1073
XM_011538838.1 XP_011537140.1 9 1411
XM_011538839.2 XP_011537141.1 9 1145
XM_011538840.2 XP_011537142.1 9 1177
XM_011538841.2 XP_011537143.1 9 1155
XM_011538842.2 XP_011537144.1 9 1135
XM_011538844.2 XP_011537146.1 9 1150
XM_011538845.1 XP_011537147.1 8 1030
XM_011538846.2 XP_011537148.1 9 1104
XM_011538847.1 XP_011537149.1 8 1005
XM_017020026.1 XP_016875515.1 9 1119
XM_017020027.1 XP_016875516.1 8 1004
GenBank mRNA AK293543.1 - 7 921
BC007277.1 - 8 1438 3343102
BC009731.1 - 8 937 3877292
BC032672.2 - 7 1559 5577070
GU211002.1 - 6 805

View Full Product Details