Gene Transcripts

Gene Symbol
BRSK1
Entrez Gene ID
84446
Gene Name
BR serine/threonine kinase 1
Gene Aliases
hSAD1
Chromosome Location
Chr. 19: 55284101 - 55312562 on Build GRCh38
UniGene ID
Hs.182081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032430.1 NP_115806.1 4 685
XM_005259327.3 XP_005259384.1 4 754
XM_011527395.2 XP_011525697.2 2 119
GenBank mRNA AB058714.1 - 3 220
AF479826.1 - 6 583
AF479827.1 - 4 685
AL834275.1 - 4 308
AY458602.1 - 4 457
AY505124.1 - 4 408
HQ830199.1 - 4 430

View Full Product Details