Gene Transcripts

Gene Symbol
KBTBD8
Entrez Gene ID
84541
Gene Name
kelch repeat and BTB domain containing 8
Gene Aliases
TA-KRP, TAKRP
Chromosome Location
Chr. 3: 66998303 - 67011210 on Build GRCh38
UniGene ID
Hs.116665
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032505.2 NP_115894.2 2 149
GenBank mRNA AF385438.1 - 1 82
AK096640.1 - 2 145
BC117487.1 - 1 46 40126038
DA246470.1 - 2 145

View Full Product Details