Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001520.2 | NP_001001520.1 | 14 | 1783 | |
NM_032631.3 | NP_116020.1 | 14 | 1783 | ||
XM_011528375.2 | XP_011526677.1 | 14 | 2143 | ||
XM_011528376.2 | XP_011526678.1 | 14 | 2140 | ||
XM_011528377.2 | XP_011526679.1 | 14 | 2143 | ||
GenBank mRNA | AB208892.1 | - | 12 | 2521 | |
AF294267.1 | - | 14 | 1690 | ||
AK027353.1 | - | 2 | 170 | ||
AY358600.1 | - | 14 | 1692 | ||
BC000755.2 | - | 14 | 1673 | 3507276 | |
BC009449.1 | - | 11 | 1322 | ||
BE745571.1 | - | 51 |