Gene Transcripts

Gene Symbol
GPAT3
Entrez Gene ID
84803
Gene Name
glycerol-3-phosphate acyltransferase 3
Gene Aliases
AGPAT 10, AGPAT10, AGPAT8, AGPAT9, HMFN0839, LPAAT-theta, MAG1
Chromosome Location
Chr. 4: 83535634 - 83605875 on Build GRCh38
UniGene ID
Hs.99196
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256421.1 NP_001243350.1 12 1367
NM_001256422.1 NP_001243351.1 12 1371
NM_032717.4 NP_116106.2 11 1454
XM_011532383.1 XP_011530685.1 10 1260
XM_011532384.2 XP_011530686.1 12 3455
XM_017008780.1 XP_016864269.1 12 1523
XM_017008781.1 XP_016864270.1 12 1530
GenBank mRNA AB075872.1 - 8 652
AK055749.1 - 12 1366
AK223543.1 - 12 1338
AY358100.1 - 11 1271
BC073136.1 - 6 430
BC090956.1 - 11 1360 30387839
DQ324782.1 - 11 1151
DQ345298.1 - 11 1451

View Full Product Details