Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032826.4 | NP_116215.1 | 10 | 1224 | |
XM_011516645.2 | XP_011514947.1 | 10 | 992 | ||
GenBank mRNA | AB052892.1 | - | 10 | 1200 | |
AJ315497.1 | - | 10 | 1056 | ||
AJ971941.1 | - | 10 | 900 | ||
AK027603.1 | - | 10 | 1064 | ||
AK075368.1 | - | 10 | 1029 | ||
AK222874.1 | - | 10 | 1073 | ||
AK307621.1 | - | 9 | 1002 | ||
BC008413.1 | - | 10 | 1045 | 4333393 |