Gene Transcripts

Gene Symbol
LTV1
Entrez Gene ID
84946
Gene Name
LTV1 ribosome biogenesis factor
Gene Aliases
C6orf93, dJ468K18.4
Chromosome Location
Chr. 6: 143843371 - 143863806 on Build GRCh38
UniGene ID
Hs.185675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032860.3 NP_116249.2 8 1153
GenBank mRNA AK027815.1 - 6 902
AK091447.1 - 3 4202
BC009855.2 - 8 1153 3926227

View Full Product Details