Gene Transcripts

Gene Symbol
MYLK2
Entrez Gene ID
85366
Gene Name
myosin light chain kinase 2
Gene Aliases
KMLC, MLCK, MLCK2, skMLCK
Chromosome Location
Chr. 20: 31819375 - 31834697 on Build GRCh38
UniGene ID
Hs.86092
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033118.3 NP_149109.1 12 1752
GenBank mRNA AF325549.1 - 11 1650
AJ272502.1 - 12 1758
BC007753.1 - 5 588
BC069627.1 - 11 1650 7262446
BC092413.1 - 12 1752 30530456
BC127622.1 - 11 1650 40085085
BX647635.1 - 12 3888

View Full Product Details