Gene Transcripts

Gene Symbol
LYRM7
Entrez Gene ID
90624
Gene Name
LYR motif containing 7
Gene Aliases
C5orf31, MC3DN8, MZM1L
Chromosome Location
Chr. 5: 131170914 - 131205426 on Build GRCh38
UniGene ID
Hs.115467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293735.1 NP_001280664.1 4 318
NM_181705.3 NP_859056.2 5 400
NR_121658.1 3 247
GenBank mRNA AK000803.1 - 5 371
BC037930.1 - 5 352 5284728
BC047079.1 - 5 330 5286889
BG714193.1 - 4 259
BI601105.1 - 5 376
BI667583.1 - 3 222
DA996312.1 - 5 402

View Full Product Details