Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001281731.1 | NP_001268660.1 | 11 | 1602 | |
NM_147190.3 | NP_671723.1 | 10 | 1401 | ||
NR_104035.1 | 10 | 2450 | |||
XM_005269220.2 | XP_005269277.1 | 10 | 1383 | ||
XM_006719687.3 | XP_006719750.1 | 11 | 1373 | ||
XM_011538975.2 | XP_011537277.1 | 11 | 1340 | ||
XM_017020202.1 | XP_016875691.1 | 11 | 1297 | ||
XM_017020203.1 | XP_016875692.1 | 12 | 1628 | ||
XM_017020205.1 | XP_016875694.1 | 11 | 1193 | ||
GenBank mRNA | AK026014.1 | - | 10 | 1207 | |
AK055740.1 | - | 1 | 1204 | ||
AK058033.1 | - | 8 | 1939 | ||
AK300937.1 | - | 11 | 1377 | ||
BC032565.1 | - | 10 | 1218 | 5580489 | |
BC033558.1 | - | 10 | 2237 |