Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033518.3 | NP_277053.2 | 16 | 1438 | |
XM_005272694.3 | XP_005272751.2 | 15 | 1428 | ||
XM_005272695.4 | XP_005272752.3 | 15 | 1430 | ||
XM_005272697.2 | XP_005272754.2 | 15 | 1381 | ||
XM_005272698.4 | XP_005272755.2 | 15 | 1319 | ||
XM_006724569.3 | XP_006724632.1 | 16 | 1438 | ||
XM_017029960.1 | XP_016885449.1 | 15 | 1350 | ||
XM_017029961.1 | XP_016885450.1 | 15 | 1322 | ||
GenBank mRNA | AF276889.1 | - | 15 | 2103 | |
AF318358.1 | - | 12 | 1549 | ||
AK094788.1 | - | 15 | 1357 | ||
BC019246.2 | - | 15 | 1288 | 3506044 | |
BC027721.2 | - | 11 | 979 |