Gene Transcripts
- Gene Symbol
- IP6K3
- Entrez Gene ID
- 117283
- Gene Name
- inositol hexakisphosphate kinase 3
- Gene Aliases
- IHPK3, INSP6K3
- Chromosome Location
- Chr. 6: 33721476 - 33746997 on Build GRCh38
- UniGene ID
- Hs.17253
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142883.1 |
NP_001136355.1
|
3, 4 |
732 |
|
NM_054111.4 |
NP_473452.2
|
2, 3 |
603 |
|
XM_005248842.3 |
XP_005248899.1
|
3, 4 |
512 |
|
XM_005248843.3 |
XP_005248900.1
|
1, 2 |
373 |
|
XM_011514295.2 |
XP_011512597.1
|
2, 3 |
615 |
|
GenBank mRNA |
AF393812.1 |
- |
1, 2 |
298 |
|
AK056586.2 |
- |
2, 3 |
523 |
|
AK223572.1 |
- |
2, 3 |
526 |
|
AK289935.1 |
- |
3, 4 |
652 |
|
BC089389.1 |
- |
2, 3 |
603 |
30530548 |
View Full Product Details