Gene Transcripts

Gene Symbol
CLRN3
Entrez Gene ID
119467
Gene Name
clarin 3
Gene Aliases
TMEM12, USH3AL1
Chromosome Location
Chr. 10: 127877841 - 127892947 on Build GRCh38
UniGene ID
Hs.242014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152311.3 NP_689524.1 1 297
XM_011539274.2 XP_011537576.1 1 296
GenBank mRNA BC029478.1 - 1 297
BM830163.1 - 1 50

View Full Product Details