Gene Transcripts

Gene Symbol
KDELC2
Entrez Gene ID
143888
Gene Name
KDEL motif containing 2
Gene Aliases
Chromosome Location
Chr. 11: 108472106 - 108498432 on Build GRCh38
UniGene ID
Hs.83286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153705.4 NP_714916.3 1, 2 271
XM_011542623.2 XP_011540925.1 1, 2 238
GenBank mRNA AF533708.1 - 1, 2 230
AK313534.1 - 1, 2 133
BC036526.2 - 1, 2 225 5266665
DN999478.1 - 1, 2 279

View Full Product Details