Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_194285.2 | NP_919261.2 | 3 | 1823 | |
XM_011519919.2 | XP_011518221.1 | 3 | 1681 | ||
GenBank mRNA | AF452716.1 | - | 1 | 1217 | |
AK096760.1 | - | 3 | 1843 | ||
AK304078.1 | - | 3 | 1739 | ||
BC036844.1 | - | 1 | 443 | ||
BX641102.1 | - | 3 | 1724 | ||
BX647798.1 | - | 3 | 1823 | ||
BX648114.1 | - | 1 | 1402 | ||
BX648478.1 | - | 3 | 1745 | ||
CR749626.1 | - | 1 | 1266 |