Gene Transcripts

Gene Symbol
AQP4-AS1
Entrez Gene ID
147429
Gene Name
AQP4 antisense RNA 1
Gene Aliases
C18orf16, CHST9-AS1
Chromosome Location
Chr. 18: 26865308 - 26935946 on Build GRCh38
UniGene ID
Hs.559280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026908.1 7 631
GenBank mRNA AK055069.1 - 7 631
BC105127.1 - 3 300
BC112070.1 - 3 300

View Full Product Details