Gene Transcripts

Gene Symbol
ZMAT2
Entrez Gene ID
153527
Gene Name
zinc finger matrin-type 2
Gene Aliases
Ptg-12, Snu23
Chromosome Location
Chr. 5: 140700360 - 140706681 on Build GRCh38
UniGene ID
Hs.350194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144723.2 NP_653324.1 2 180
GenBank mRNA AK055683.1 - 2 93
BC056668.1 - 2 97 4773709
BP210985.1 - 2 174

View Full Product Details