Gene Transcripts

Gene Symbol
VPS13B
Entrez Gene ID
157680
Gene Name
vacuolar protein sorting 13 homolog B
Gene Aliases
CHS1, COH1
Chromosome Location
Chr. 8: 99007014 - 99877586 on Build GRCh38
UniGene ID
Hs.191540
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015243.2 NP_056058.2 2, 3 253
NM_017890.4 NP_060360.3 2, 3 253
NM_152564.4 NP_689777.3 2, 3 253
NM_181661.2 NP_858047.2 2, 3 253
NR_047582.1 2, 3 253
XM_005250800.3 XP_005250857.1 2, 3 559
XM_005250801.4 XP_005250858.1 2, 3 6429
XM_006716510.3 XP_006716573.1 2, 3 245
XM_011516848.2 XP_011515150.1 2, 3 245
XM_011516849.2 XP_011515151.1 2, 3 245
XM_011516853.2 XP_011515155.1 2, 3 245
XM_011516859.2 XP_011515161.1 2, 3 245
XM_011516861.1 XP_011515163.1 2, 3 245
XM_011516865.1 XP_011515167.1 2, 3 245
XM_011516866.2 XP_011515168.1 2, 3 245
XM_017013109.1 XP_016868598.1 2, 3 245
GenBank mRNA AJ608772.1 - 2, 3 245
AJ608773.1 - 2, 3 245
AK000590.1 - 2, 3 279
AK223122.1 - 2, 3 279
AY223814.1 - 2, 3 253
AY223815.1 - 2, 3 253
AY223816.1 - 2, 3 253
AY223817.1 - 2, 3 253
AY223818.1 - 2, 3 253
BC094749.1 - 2, 3 209
BC132976.1 - 1, 2 176
BC132978.1 - 1, 2 176

View Full Product Details