Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144071.1 | NP_001137543.1 | 8 | 1128 | |
NM_144972.4 | NP_659409.2 | 7 | 1721 | ||
XM_005252805.4 | XP_005252862.2 | 8 | 1010 | ||
XM_011519922.2 | XP_011518224.1 | 9 | 1066 | ||
XM_011519923.2 | XP_011518225.1 | 9 | 1153 | ||
XM_011519924.2 | XP_011518226.1 | 9 | 1010 | ||
XM_011519925.1 | XP_011518227.1 | 9 | 992 | ||
GenBank mRNA | AK131523.1 | - | 7 | 1664 | |
AY581313.1 | - | 8 | 1128 |