Gene Transcripts

Gene Symbol
FAM71D
Entrez Gene ID
161142
Gene Name
family with sequence similarity 71 member D
Gene Aliases
C14orf54
Chromosome Location
Chr. 14: 67188365 - 67228550 on Build GRCh38
UniGene ID
Hs.412818
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173526.3 NP_775797.2 8 1497
XM_011536500.1 XP_011534802.1 8 1461
XM_011536502.1 XP_011534804.1 8 1396
XM_011536503.2 XP_011534805.1 7 2438
GenBank mRNA AK093449.1 - 8 1461
BC050401.1 - 9 1363 5745029

View Full Product Details