Gene Transcripts

Gene Symbol
EXD1
Entrez Gene ID
161829
Gene Name
exonuclease 3'-5' domain containing 1
Gene Aliases
EXDL1
Chromosome Location
Chr. 15: 41182728 - 41231074 on Build GRCh38
UniGene ID
Hs.307999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152596.3 NP_689809.2 1, 2 270
GenBank mRNA AK292524.1 - 1, 2 176
BC030628.2 - 1, 2 210 4827307

View Full Product Details