Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040425.2 | NP_001035515.1 | 6 | 555 | |
NM_144987.3 | NP_659424.2 | 6 | 497 | ||
NR_110171.1 | 8 | 834 | |||
NR_110172.1 | 6 | 762 | |||
NR_110173.1 | 7 | 610 | |||
GenBank mRNA | AI767079.1 | - | 1 | 241 | |
AK092844.1 | - | 3 | 1532 | ||
AK094213.1 | - | 3 | 2352 | ||
AY461582.1 | - | 8 | 808 | ||
AY569437.3 | - | 6 | 524 | ||
BC021186.1 | - | 6 | 466 | 5260387 | |
BM696851.1 | - | 7 | 603 | ||
CD175846.1 | - | 6 | 506 |