Gene Transcripts

Gene Symbol
KCTD6
Entrez Gene ID
200845
Gene Name
potassium channel tetramerization domain containing 6
Gene Aliases
KCASH3
Chromosome Location
Chr. 3: 58492096 - 58502360 on Build GRCh38
UniGene ID
Hs.13982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128214.1 NP_001121686.1 3 760
NM_153331.3 NP_699162.3 2 1034
XM_005264937.2 XP_005264994.1 3 2092
GenBank mRNA AK027572.1 - 2 1626
AK074934.1 - 2 1034
AK075297.1 - 3 742
BC022893.1 - 2 776 4689885

View Full Product Details