Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290341.2 | NP_001277270.1 | 10 | 1270 | |
NM_001290342.2 | NP_001277271.1 | 9 | 1096 | ||
NM_001290343.1 | NP_001277272.1 | 9 | 1007 | ||
NM_178842.4 | NP_849164.2 | 9 | 1003 | ||
XM_011521355.2 | XP_011519657.1 | 10 | 1313 | ||
XM_011521357.2 | XP_011519659.1 | 10 | 1207 | ||
XM_011521358.1 | XP_011519660.1 | 10 | 1091 | ||
XM_017022002.1 | XP_016877491.1 | 9 | 1087 | ||
XM_017022003.1 | XP_016877492.1 | 9 | 1012 | ||
XM_017022004.1 | XP_016877493.1 | 8 | 923 | ||
GenBank mRNA | BC028703.1 | - | 9 | 1084 | |
BC034500.1 | - | 9 | 959 | 4838423 | |
BC034970.1 | - | 9 | 1005 | 4822006 |