Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039547.2 | NP_001034636.1 | 2 | 339 | |
NR_033289.1 | 2 | 339 | |||
XM_011512624.2 | XP_011510926.1 | 2 | 137 | ||
XM_017006088.1 | XP_016861577.1 | 2 | 324 | ||
XM_017006089.1 | XP_016861578.1 | 2 | 307 | ||
XM_017006090.1 | XP_016861579.1 | 2 | 324 | ||
GenBank mRNA | AK127641.1 | - | 2 | 307 | |
AK310761.1 | - | 2 | 324 | ||
AK313792.1 | - | 2 | 221 | ||
BC032470.1 | - | 2 | 299 | 5217372 | |
BX648359.1 | - | 2 | 319 | ||
BX648681.1 | - | 2 | 280 | ||
DR000158.1 | - | 2 | 340 |