Gene Transcripts

Gene Symbol
C10orf53
Entrez Gene ID
282966
Gene Name
chromosome 10 open reading frame 53
Gene Aliases
Chromosome Location
Chr. 10: 49679638 - 49708910 on Build GRCh38
UniGene ID
Hs.131287
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042427.1 NP_001035892.1 2 199
NM_182554.2 NP_872360.2 2 199
GenBank mRNA AK091773.1 - 2 186
BC028127.1 - 2 151 5164374
DB043416.1 - 2 172

View Full Product Details