Gene Transcripts

Gene Symbol
UBAC2
Entrez Gene ID
337867
Gene Name
UBA domain containing 2
Gene Aliases
PHGDHL1
Chromosome Location
Chr. 13: 99200425 - 99386499 on Build GRCh38
UniGene ID
Hs.508545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144072.1 NP_001137544.1 5 929
NM_177967.3 NP_808882.1 3 843
NR_026644.1 5 1073
XM_006719948.3 XP_006720011.1 6 639
XM_011521082.2 XP_011519384.1 3 699
XM_011521083.2 XP_011519385.1 4 406
XM_011521084.2 XP_011519386.1 5 494
GenBank mRNA AK054563.1 - 3 844
AK075419.1 - 5 500
AK075516.1 - 3 270
AK129861.1 - 5 528
AK290342.1 - 5 929
AK300908.1 - 5 415
BC063559.1 - 3 288
BC121139.1 - 5 672 40120987

View Full Product Details