Gene Transcripts
- Gene Symbol
- OR52A4P
- Entrez Gene ID
- 390053
- Gene Name
- olfactory receptor family 52 subfamily A member 4 pseudogene
- Gene Aliases
- OR52A4
- Chromosome Location
- Chr. 11: 5120664 - 5121578 on Build GRCh38
- UniGene ID
- Hs.553729
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC140752.1 |
- |
1 |
251 |
|
View Full Product Details