Gene Transcripts

Gene Symbol
OR6C1
Entrez Gene ID
390321
Gene Name
olfactory receptor family 6 subfamily C member 1
Gene Aliases
OST267
Chromosome Location
Chr. 12: 55320600 - 55321538 on Build GRCh38
UniGene ID
Hs.553763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005182.1 NP_001005182.1 1 231
GenBank mRNA BC136957.1 - 1 270
BC136965.1 - 1 269 9020957

View Full Product Details