Gene Transcripts

Gene Symbol
ATP2A3
Entrez Gene ID
489
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 3
Gene Aliases
SERCA3
Chromosome Location
Chr. 17: 3923869 - 3964464 on Build GRCh38
UniGene ID
Hs.513870
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005173.3 NP_005164.2 15 2438
NM_174953.2 NP_777613.1 15 2438
NM_174954.2 NP_777614.1 15 2438
NM_174955.2 NP_777615.1 15 2438
NM_174956.2 NP_777616.1 15 2438
NM_174957.2 NP_777617.1 15 2438
NM_174958.2 NP_777618.1 15 2438
XM_011523881.2 XP_011522183.1 15 2438
XM_011523882.2 XP_011522184.1 15 2438
XM_011523884.2 XP_011522186.1 15 2438
XM_011523885.1 XP_011522187.1 15 2438
XM_011523888.2 XP_011522190.1 15 2438
XM_011523889.1 XP_011522191.1 15 2438
XM_017024692.1 XP_016880181.1 15 2438
XM_017024693.1 XP_016880182.1 13 2064
GenBank mRNA AF068220.1 - 15 2389
AF068221.1 - 3 666
AK292716.1 - 15 2438
BC035729.1 - 15 2393 5723849
Z69881.1 - 15 2271

View Full Product Details