Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021077.3 | NP_066563.2 | 1 | 455 | |
NM_205858.1 | NP_995580.1 | 1 | 455 | ||
XM_017022239.1 | XP_016877728.1 | 1 | 495 | ||
GenBank mRNA | AL110112.1 | - | 1 | 74 | |
BC007407.2 | - | 1 | 448 | 3030430 | |
BC007431.2 | - | 1 | 448 | 3030430 | |
BC008603.1 | - | 1 | 75 | 4185578 | |
BI858080.1 | - | 1 | 408 | ||
BI962836.1 | - | 1 | 455 | ||
M21551.1 | - | 1 | 72 |