Gene Transcripts
- Gene Symbol
- RAB5C
- Entrez Gene ID
- 5878
- Gene Name
- RAB5C, member RAS oncogene family
- Gene Aliases
- L1880, RAB5CL, RAB5L, RABL
- Chromosome Location
- Chr. 17: 42124976 - 42155044 on Build GRCh38
- UniGene ID
- Hs.650382
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252039.1 |
NP_001238968.1
|
4, 5 |
783 |
|
NM_004583.3 |
NP_004574.2
|
3, 4 |
567 |
|
NM_201434.2 |
NP_958842.1
|
4, 5 |
684 |
|
XM_011525090.1 |
XP_011523392.1
|
3, 4 |
506 |
|
XM_011525091.1 |
XP_011523393.1
|
3, 4 |
630 |
|
GenBank mRNA |
AF141304.1 |
- |
2, 3 |
396 |
|
AF498938.1 |
- |
2, 3 |
327 |
|
AK310234.1 |
- |
4, 5 |
727 |
|
AL553867.3 |
- |
3, 4 |
522 |
|
BC106039.1 |
- |
3, 4 |
526 |
4938169 |
BC114439.1 |
- |
3, 4 |
522 |
40083572 |
BM424166.1 |
- |
4, 5 |
624 |
|
BM550607.1 |
- |
3, 4 |
527 |
|
BT019484.1 |
- |
2, 3 |
327 |
|
CR541901.1 |
- |
2, 3 |
327 |
|
U11293.1 |
- |
2, 3 |
402 |
|
U18420.1 |
- |
3, 4 |
462 |
|
View Full Product Details