Gene Transcripts
- Gene Symbol
- CCL21
- Entrez Gene ID
- 6366
- Gene Name
- C-C motif chemokine ligand 21
- Gene Aliases
- 6Ckine, CKb9, ECL, SCYA21, SLC, TCA4
- Chromosome Location
- Chr. 9: 34709005 - 34710167 on Build GRCh38
- UniGene ID
- Hs.57907
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002989.3 |
NP_002980.1
|
2, 3 |
278 |
|
XM_011518004.2 |
XP_011516306.1
|
2, 3 |
265 |
|
GenBank mRNA |
AB002409.1 |
- |
2, 3 |
235 |
|
AF001979.1 |
- |
2, 3 |
177 |
|
AK311901.1 |
- |
2, 3 |
247 |
|
AY358887.1 |
- |
2, 3 |
182 |
|
BC027918.1 |
- |
2, 3 |
184 |
5226781 |
BI833188.1 |
- |
2, 3 |
260 |
|
BP331359.1 |
- |
2, 3 |
278 |
|
CR450326.1 |
- |
2, 3 |
177 |
|
U88320.1 |
- |
2, 3 |
191 |
|
View Full Product Details