Gene Transcripts
- Gene Symbol
- TNFRSF4
- Entrez Gene ID
- 7293
- Gene Name
- TNF receptor superfamily member 4
- Gene Aliases
- ACT35, CD134, IMD16, OX40, TXGP1L
- Chromosome Location
- Chr. 1: 1211326 - 1216812 on Build GRCh38
- UniGene ID
- Hs.129780
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003327.3 |
NP_003318.1
|
4, 5 |
472 |
|
XM_011542074.2 |
XP_011540376.1
|
2, 3 |
826 |
|
XM_011542075.2 |
XP_011540377.1
|
4, 5 |
939 |
|
XM_011542076.2 |
XP_011540378.1
|
2, 3 |
826 |
|
XM_011542077.2 |
XP_011540379.1
|
2, 3 |
185 |
|
XM_017002231.1 |
XP_016857720.1
|
5, 6 |
1817 |
|
XM_017002232.1 |
XP_016857721.1
|
5, 6 |
1817 |
|
GenBank mRNA |
BC105070.1 |
- |
4, 5 |
472 |
|
BC105072.1 |
- |
4, 5 |
472 |
8144075 |
S76792.1 |
- |
4, 5 |
422 |
|
View Full Product Details