Gene Transcripts

Gene Symbol
PEX16
Entrez Gene ID
9409
Gene Name
peroxisomal biogenesis factor 16
Gene Aliases
PBD8A, PBD8B
Chromosome Location
Chr. 11: 45909669 - 45918123 on Build GRCh38
UniGene ID
Hs.100915
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004813.2 NP_004804.1 3 510
NM_057174.2 NP_476515.1 3 510
XM_011520474.1 XP_011518776.1 3 249
GenBank mRNA AB016531.1 - 3 235
AF118240.1 - 3 442
BC000467.2 - 3 216 2822827
BC004356.1 - 3 208 2822311
CN285513.1 - 3 517

View Full Product Details