Gene Transcripts

Gene Symbol
TMEM63A
Entrez Gene ID
9725
Gene Name
transmembrane protein 63A
Gene Aliases
KIAA0792
Chromosome Location
Chr. 1: 225840556 - 225882720 on Build GRCh38
UniGene ID
Hs.119387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014698.2 NP_055513.2 12, 13 1575
XM_006711841.3 XP_006711904.1 10, 11 1068
XM_011544328.2 XP_011542630.1 12, 13 1427
XM_011544329.2 XP_011542631.1 12, 13 2694
XM_011544330.2 XP_011542632.1 11, 12 1384
XM_011544331.2 XP_011542633.1 12, 13 1258
XM_011544332.2 XP_011542634.1 11, 12 1206
XM_017002916.1 XP_016858405.1 11, 12 1658
GenBank mRNA AB007958.1 - 3, 4 2720
AB018335.1 - 12, 13 1203
AK022502.1 - 12, 13 1224
AK222934.1 - 12, 13 1205
AK222944.1 - 12, 13 1205
BC030245.1 - 12, 13 1208 5216414
BC113811.1 - 5, 6 616

View Full Product Details