Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098482.1 | NP_001091952.1 | 2 | 286 | |
NM_015321.2 | NP_056136.2 | 2 | 286 | ||
XM_005259833.3 | XP_005259890.1 | 2 | 247 | ||
XM_005259834.1 | XP_005259891.1 | 2 | 280 | ||
XM_005259835.3 | XP_005259892.1 | 2 | 246 | ||
XM_005259836.3 | XP_005259893.1 | 2 | 246 | ||
XM_006722710.3 | XP_006722773.1 | 2 | 246 | ||
XM_011527842.2 | XP_011526144.1 | 2 | 247 | ||
XM_017026536.1 | XP_016882025.1 | 2 | 247 | ||
XM_017026537.1 | XP_016882026.1 | 2 | 247 | ||
XM_017026538.1 | XP_016882027.1 | 2 | 247 | ||
GenBank mRNA | AB014516.1 | - | 2 | 151 | |
AK024089.1 | - | 2 | 286 | ||
AU118537.1 | - | 2 | 286 | ||
AY040323.1 | - | 2 | 286 | ||
AY186998.1 | - | 172 | |||
AY360171.1 | - | 2 | 223 | ||
BC023614.2 | - | 2 | 109 | ||
BC028050.1 | - | 2 | 218 | 4938995 |