Gene Transcripts
- Gene Symbol
- PPY2P
- Entrez Gene ID
- 23614
- Gene Name
- pancreatic polypeptide 2, pseudogene
- Gene Aliases
- PPY2
- Chromosome Location
- Chr. 17: 28247444 - 28248289 on Build GRCh38
- UniGene ID
- Hs.744962
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002181.1 |
|
1 |
149 |
|
GenBank mRNA |
AF085945.1 |
- |
1 |
44 |
|
AF222903.1 |
- |
1 |
149 |
|
- Gene Symbol
- LOC105371712
- Entrez Gene ID
- 105371712
- Gene Name
- uncharacterized LOC105371712
- Gene Aliases
- Chromosome Location
- Chr. 17: 28246454 - 28250825 on Build GRCh38
- UniGene ID
- Hs.20588
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC082966.1 |
- |
1 |
411 |
|
View Full Product Details