Gene Transcripts

Gene Symbol
HYALP1
Entrez Gene ID
26062
Gene Name
hyaluronoglucosaminidase pseudogene 1
Gene Aliases
HYAL6
Chromosome Location
Chr. 7: 123814139 - 123819430 on Build GRCh38
UniGene ID
Hs.381305
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002731.1
1 248
GenBank mRNA AF149304.1 - LIT2068
HYALP1
1 248

View Full Product Details