Gene Transcripts

Gene Symbol
YBEY
Entrez Gene ID
54059
Gene Name
ybeY metallopeptidase (putative)
Gene Aliases
C21orf57
Chromosome Location
Chr. 21: 46286056 - 46314188 on Build GRCh38
UniGene ID
Hs.474066
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006114.2 NP_001006114.1 3 425
NM_001314023.1 NP_001300952.1 3 290
NM_001314025.1 NP_001300954.1 3 425
NM_001314026.1 NP_001300955.1 3 747
NM_058181.2 NP_478061.1 3 747
XM_011529629.2 XP_011527931.1 3 395
XM_011529630.2 XP_011527932.1 3 426
XM_011529631.2 XP_011527933.1 3 395
XM_011529633.2 XP_011527935.1 3 395
XM_011529635.2 XP_011527937.1 3 426
XM_017028396.1 XP_016883885.1 442
GenBank mRNA AI262212.1 - 3 302
AK128875.1 - 3 359
AK294975.1 - 3 406
AY040873.1 - 3 693
AY040875.1 - 3 236
BC022828.1 - 3 366 3928883
BC119770.1 - 3 371 40108760
BC122865.1 - 3 371 40108759
BC131553.1 - 3 371 40089560
BG473705.1 - 2 331
BM796288.1 - 3 418

View Full Product Details