Gene Transcripts

Gene Symbol
ANKIB1
Entrez Gene ID
54467
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
Chromosome Location
Chr. 7: 92246216 - 92401773 on Build GRCh38
UniGene ID
Hs.83293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019004.1 NP_061877.1 20 3206
XM_005250458.3 XP_005250515.1 20 3206
XM_011516334.1 XP_011514636.1 19 3041
XM_011516335.2 XP_011514637.1 20 2788
XM_017012359.1 XP_016867848.1 18 3524
XM_017012360.1 XP_016867849.1 16 1999
GenBank mRNA AB037807.1 - 20 3206
AK001179.1 - 9 1213
AK025475.1 - 17 2303
AL137349.1 - 9 1188
BC031483.1 - 13 1735
BC063861.1 - 20 3182
BC073893.1 - 14 1798
BC094865.1 - 6 888

View Full Product Details