Gene Transcripts

Gene Symbol
DOCK10
Entrez Gene ID
55619
Gene Name
dedicator of cytokinesis 10
Gene Aliases
DRIP2, Nbla10300, ZIZ3
Chromosome Location
Chr. 2: 224765090 - 225042689 on Build GRCh38
UniGene ID
Hs.46578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290263.1 NP_001277192.1 32 3521
NM_014689.2 NP_055504.2 32 3664
XM_005246673.4 XP_005246730.1 32 3523
XM_006712617.3 XP_006712680.1 32 3688
XM_017004441.1 XP_016859930.1 32 3691
XM_017004442.1 XP_016859931.1 32 3694
XM_017004443.1 XP_016859932.1 32 3740
XM_017004444.1 XP_016859933.1 31 3647
XM_017004445.1 XP_016859934.1 32 3521
XM_017004446.1 XP_016859935.1 31 3545
XM_017004447.1 XP_016859936.1 32 3690
GenBank mRNA AK293879.1 - 3 97
CR749492.1 - 7 509
EU236710.2 - 32 3518

View Full Product Details