Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024660.2 | NP_078936.1 | 4 | 628 | |
GenBank mRNA | AK026226.1 | - | 4 | 629 | |
AX775837.1 | - | 4 | 631 | ||
BC126481.1 | - | 4 | 589 | 8992197 | |
BC126483.1 | - | 4 | 589 | 8992199 | |
BC144205.1 | - | 4 | 587 |