Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136201.1 | NP_001129673.1 | 2 | 249 | |
NM_001136202.1 | NP_001129674.1 | 2 | 249 | ||
NM_024710.2 | NP_078986.1 | 2 | 249 | ||
GenBank mRNA | AK027122.1 | - | 2 | 212 | |
AK131324.1 | - | 2 | 910 | ||
AK311540.1 | - | 199 | |||
BC017344.2 | - | 2 | 136 | 4893552 | |
BP226043.1 | - | 2 | 250 | ||
CR457377.1 | - | 1 | 75 |