Gene Transcripts
- Gene Symbol
- C15orf48
- Entrez Gene ID
- 84419
- Gene Name
- chromosome 15 open reading frame 48
- Gene Aliases
- FOAP-11, NMES1
- Chromosome Location
- Chr. 15: 45430529 - 45433449 on Build GRCh38
- UniGene ID
- Hs.112242
- Species
- Homo sapiens
- Gene Symbol
- MIR147B
- Entrez Gene ID
- 100126311
- Gene Name
- microRNA 147b
- Gene Aliases
- MIRN147B, mir-147b
- Chromosome Location
- Chr. 15: 45433050 - 45433129 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030599.1 |
|
1 |
56 |
|
View Full Product Details