Gene Transcripts

Gene Symbol
SNORD15B
Entrez Gene ID
114599
Gene Name
small nucleolar RNA, C/D box 15B
Gene Aliases
RNU15B, U15B
Chromosome Location
Chr. 11: 75404421 - 75404566 on Build GRCh38
UniGene ID
Hs.689080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_000025.1 1 101
GenBank mRNA L15557.1 - 1 101

View Full Product Details