Gene Transcripts

Gene Symbol
VWA3A
Entrez Gene ID
146177
Gene Name
von Willebrand factor A domain containing 3A
Gene Aliases
Chromosome Location
Chr. 16: 22091930 - 22156966 on Build GRCh38
UniGene ID
Hs.10697
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173615.4 NP_775886.3 30, 31 3390
XM_011545742.2 XP_011544044.1 34, 35 3672
XM_011545744.2 XP_011544046.1 19, 20 2272
XM_011545745.1 XP_011544047.1 17, 18 2040
XM_011545746.2 XP_011544048.1 17, 18 2143
XM_017022947.1 XP_016878436.1 35, 36 3745
XM_017022948.1 XP_016878437.1 18, 19 2844
XM_017022949.1 XP_016878438.1 16, 17 3115
GenBank mRNA AK093467.1 - 6, 7 953
AK128606.1 - 19, 20 2258
AY360464.1 - 5, 6 596
BC038400.1 - 7, 8 835
BC109294.1 - 19, 20 2165
BC109295.1 - 18, 19 2118

View Full Product Details